Learn More
Sampling Systems™ Double-ended Spoon
These double-ended spoons are ideal for taking different sample sizes using the same spoon. The lengths and volume capacities of the spoons vary, allowing for flexibility in sampling both powders and liquids.
Marke: Sampling Systems™ A104-300
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Made from stainless steel
- Ideal for sampling powders and liquids
- Various sizes available
Spezifikation
Löffel | |
Autoklavierbar | |
No | |
300 mm |
Stainless Steel | |
Reusable | |
Unsteril |