Learn More
Tocris Bioscience™ DL-AP5 Sodium salt
Potent and selective NMDA antagonist; sodium salt of DL-AP5
Marke: Tocris Bioscience™ 3693/50
363.87 EUR gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Beschreibung
DL-AP5 Sodium salt is the sodium salt of DL-AP5 (Cat. No. 0105) and a racemic mixture of the D- and L-isomers of AP5, a selective NMDA receptor antagonist that competes with glutamate binding and is commonly used to inhibit NMDA-dependent synaptic plasticity. D-AP5 (Cat. No. 0106) is the more active isomer and displays approximately 52-fold higher potency than the L-isomer, L-AP5 (Cat. No. 0107). In vitro D-AP5 reduces NMDA-induced depolarization of cortical neurons, with no effect on the response to L-Quisqualic acid (Cat. No. 0188) or Kainic acid (Cat. No. 0222). Following spinal injection of D-AP5, NMDA-response is rapidly reduced, with no effect seen on spontaneously active neurons.DL mixture, D-isomer and L-isomer also available.
Spezifikation
>98% | |
DL-AP5 Sodium salt | |
DL-AP5 Sodium salt |
50 mg | |
219.11 |
Nur für Forschungszwecke