Learn More
Tuttnauer 21 CFR Part 11 Software
Record of performed cycles and of actions taken by operator using software and system for operating the autoclave. R.PC.R is required for 21 CFR part 11 in order to save a history log of cycles and track changes made to the autoclave parameters by operators.
Marke:  Tuttnauer ADD222-0442
Artikelnummer. 17829558
  • 2530.00 € / 1 Stück
Verfügbar ab: 21-05-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
  • Record of performed cycles.
  • Record of actions taken by operator using software and system for operating the autoclave (according to user access levels).
  • This information can be used for auditing the operation of the autoclave.
  • R.PC.R is required for 21 CFR part 11 in order to save a history log of cycles and track changes made to the autoclave parameters by operators.
  • 21 CFR part 11 - Determined by the manufacturers policy.
Spezifikation
Spezifikation
21 CFR Part 11
Software
Dokumentation
One moment while we fetch your results.
Name des Produkts
Tuttnauer 21 CFR Part 11 Software > 21 CFR Part 11 Software