missing translation for 'onlineSavingsMsg'
Learn More

2-[4-Fluoro-3-(trifluoromethyl)phenyl]ethanol, 90+%, Thermo Scientific™

Product Code. 15468225
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15468225

Brand: Thermo Scientific Alfa Aesar H32867.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 618 41 11 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886761-80-4
Molecular Formula C9H8F4O
Molecular Weight (g/mol) 208.16
MDL Number MFCD06660359
InChI Key WNWSWMPFXNNAQT-UHFFFAOYSA-N
Synonym 2-4-fluoro-3-trifluoromethyl phenyl ethanol, 2-4-fluoro-3-trifluoromethyl phenyl ethan-1-ol
PubChem CID 17999363
IUPAC Name 2-[4-fluoro-3-(trifluoromethyl)phenyl]ethanol
SMILES OCCC1=CC(=C(F)C=C1)C(F)(F)F

Specifications

Refractive Index 1.4565
Quantity 1 g
Formula Weight 208.15
Percent Purity ≥90%
Chemical Name or Material 2-[4-Fluoro-3-(trifluoromethyl)phenyl]ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.