CAS RN 1053658-84-6
CAS RN 1053658-84-6
IUPAC Name:
(2-aminoethyl)di-tert-butyl)phosphane
Synonyme:
(2-aminoethyl)di-tert-butyl)phosphane
Molekulargewicht (g/mol):
189.28
Summenformel:
C10H24NP
InChi Key:
DZDGRVHIUYIMDS-UHFFFAOYSA-N
SMILES:
CC(C)(C)P(CCN)C(C)(C)C