CAS RN 103890-78-4
CAS RN 103890-78-4
IUPAC Name:
3,5-diethyl 4-{2-[(1E)-3-(tert-butoxy)-3-oxoprop-1-en-1-yl]phenyl}-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Synonyme:
lacidipine glaxosmithkline brand of lacidipine lacidipine (product) lacidipinum lacidipino motens boehringer ingelheim brand of lacidipine menarini brand of lacidipine 拉西地平 lacipil
Molekulargewicht (g/mol):
455.55
Summenformel:
C26H33NO6
InChi Key:
GKQPCPXONLDCMU-CCEZHUSRSA-N
SMILES:
CCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1\C=C\C(=O)OC(C)(C)C)C(=O)OCC