CAS RN 10377-51-2
CAS RN 10377-51-2
Lithiumiodid, 99 %, rein, wasserfrei, Thermo Scientific Chemicals
CAS: 10377-51-2 Summenformel: ILi Molekulargewicht (g/mol): 133.84 MDL-Nummer: MFCD00011092 InChI-Schlüssel: HSZCZNFXUDYRKD-UHFFFAOYSA-M Synonym: lithium iodide,lithium iodide, anhydrous,unii-s6k2xet783,lithium iodide 6lii,lithium 1+ ion iodide,lithium iodide, ultra dry,lithium iodide 7lii,lithiumiodide,lithium monoiodide PubChem CID: 66321 IUPAC-Name: Lithium;iodid SMILES: [Li+].[I-]
Lithiumiodid, ultratrocken, 99.999 % (Metallbasis), Thermo Scientific Chemicals
CAS: 10377-51-2 Summenformel: ILi Molekulargewicht (g/mol): 133.84 MDL-Nummer: MFCD00011092 InChI-Schlüssel: HSZCZNFXUDYRKD-UHFFFAOYSA-M Synonym: lithium iodide,lithium iodide, anhydrous,unii-s6k2xet783,lithium iodide 6lii,lithium 1+ ion iodide,lithium iodide, ultra dry,lithium iodide 7lii,lithiumiodide,lithium monoiodide PubChem CID: 66321 IUPAC-Name: Lithium;iodid SMILES: [Li+].[I-]
Lithiumiodid, wasserfrei, 98+ %, Thermo Scientific Chemicals
CAS: 10377-51-2 Summenformel: ILi Molekulargewicht (g/mol): 133.84 MDL-Nummer: MFCD00011092 InChI-Schlüssel: HSZCZNFXUDYRKD-UHFFFAOYSA-M Synonym: lithium iodide,lithium iodide, anhydrous,unii-s6k2xet783,lithium iodide 6lii,lithium 1+ ion iodide,lithium iodide, ultra dry,lithium iodide 7lii,lithiumiodide,lithium monoiodide PubChem CID: 66321 IUPAC-Name: Lithium;iodid SMILES: [Li+].[I-]
Lithiumiodid, ultratrocken, 99 % (Metallbasis), Thermo Scientific Chemicals
CAS: 10377-51-2 Summenformel: ILi Molekulargewicht (g/mol): 133.84 MDL-Nummer: MFCD00011092 InChI-Schlüssel: HSZCZNFXUDYRKD-UHFFFAOYSA-M Synonym: lithium iodide,lithium iodide, anhydrous,unii-s6k2xet783,lithium iodide 6lii,lithium 1+ ion iodide,lithium iodide, ultra dry,lithium iodide 7lii,lithiumiodide,lithium monoiodide PubChem CID: 66321 IUPAC-Name: Lithium;iodid SMILES: [Li+].[I-]
Lithiumiodid, wasserfrei, 99.95 % (Metallbasis), Thermo Scientific Chemicals
CAS: 10377-51-2 Summenformel: ILi Molekulargewicht (g/mol): 133.84 MDL-Nummer: MFCD00011092 InChI-Schlüssel: HSZCZNFXUDYRKD-UHFFFAOYSA-M Synonym: lithium iodide,lithium iodide, anhydrous,unii-s6k2xet783,lithium iodide 6lii,lithium 1+ ion iodide,lithium iodide, ultra dry,lithium iodide 7lii,lithiumiodide,lithium monoiodide PubChem CID: 66321 IUPAC-Name: Lithium;iodid SMILES: [Li+].[I-]
Lithiumiodid, ultratrocken, weißes Pulver, 99.999 % (Metallbasis), Thermo Scientific Chemicals
CAS: 10377-51-2 Summenformel: ILi Molekulargewicht (g/mol): 133.84 MDL-Nummer: MFCD00011092 InChI-Schlüssel: HSZCZNFXUDYRKD-UHFFFAOYSA-M Synonym: lithium iodide,lithium iodide, anhydrous,unii-s6k2xet783,lithium iodide 6lii,lithium 1+ ion iodide,lithium iodide, ultra dry,lithium iodide 7lii,lithiumiodide,lithium monoiodide PubChem CID: 66321 SMILES: [Li+].[I-]