CAS RN 10101-97-0
CAS RN 10101-97-0
Nickel(II)-sulfat-Hexahydrat, 98 %, Thermo Scientific Chemicals
CAS: 10101-97-0 Summenformel: H12NiO10S Molekulargewicht (g/mol): 262.839 MDL-Nummer: MFCD00149813 InChI-Schlüssel: RRIWRJBSCGCBID-UHFFFAOYSA-L Synonym: nickel sulfate hexahydrate,nickel ii sulfate hexahydrate,nickel 2+ sulfate hexahydrate,nickel ii sulfate,nickelsulfatehexahydrate,dsstox_cid_928,acmc-1c7xm,dsstox_rid_75872,ni.so4.6h2o,dsstox_gsid_20928 PubChem CID: 5284429 ChEBI: CHEBI:53437 IUPAC-Name: Nickel(2+); Sulfat; Hexahydrat SMILES: O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
Nickel(II)-sulfat-Hexahydrat, 98+ %, ACS-Reagenz, Thermo Scientific Chemicals
CAS: 10101-97-0 Summenformel: NiO4S·6H2O Molekulargewicht (g/mol): 262.85 MDL-Nummer: MFCD00149813 InChI-Schlüssel: RRIWRJBSCGCBID-UHFFFAOYSA-L Synonym: nickel sulfate hexahydrate,nickel ii sulfate hexahydrate,nickel 2+ sulfate hexahydrate,nickel ii sulfate,nickelsulfatehexahydrate,dsstox_cid_928,acmc-1c7xm,dsstox_rid_75872,ni.so4.6h2o,dsstox_gsid_20928 PubChem CID: 5284429 ChEBI: CHEBI:53437 IUPAC-Name: Nickel(2+); Sulfat; Hexahydrat SMILES: O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
Nickel(II)-sulfat- Hexahydrat, 99 %, für Analysen, Thermo Scientific Chemicals
CAS: 10101-97-0 Summenformel: NiO4S·6H2O Molekulargewicht (g/mol): 262.85 MDL-Nummer: MFCD00149813 InChI-Schlüssel: RRIWRJBSCGCBID-UHFFFAOYSA-L Synonym: nickel sulfate hexahydrate,nickel ii sulfate hexahydrate,nickel 2+ sulfate hexahydrate,nickel ii sulfate,nickelsulfatehexahydrate,dsstox_cid_928,acmc-1c7xm,dsstox_rid_75872,ni.so4.6h2o,dsstox_gsid_20928 PubChem CID: 5284429 ChEBI: CHEBI:53437 IUPAC-Name: Nickel(2+); Sulfat; Hexahydrat SMILES: O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
Nickel(II)-sulfat-Hexahydrat, ACS, 98.0 % min., Thermo Scientific Chemicals
CAS: 10101-97-0 Summenformel: H12NiO10S Molekulargewicht (g/mol): 262.839 MDL-Nummer: MFCD00149813 InChI-Schlüssel: RRIWRJBSCGCBID-UHFFFAOYSA-L Synonym: nickel sulfate hexahydrate,nickel ii sulfate hexahydrate,nickel 2+ sulfate hexahydrate,nickel ii sulfate,nickelsulfatehexahydrate,dsstox_cid_928,acmc-1c7xm,dsstox_rid_75872,ni.so4.6h2o,dsstox_gsid_20928 PubChem CID: 5284429 ChEBI: CHEBI:53437 IUPAC-Name: Nickel(2+); Sulfat; Hexahydrat SMILES: O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
Nickel(II)-sulfat, 0.5 M wässrige Lösung, Thermo Scientific Chemicals
CAS: 10101-97-0 Summenformel: H12NiO10S Molekulargewicht (g/mol): 262.839 MDL-Nummer: MFCD00011146 InChI-Schlüssel: RRIWRJBSCGCBID-UHFFFAOYSA-L Synonym: nickel sulfate hexahydrate,nickel ii sulfate hexahydrate,nickel 2+ sulfate hexahydrate,nickel ii sulfate,nickelsulfatehexahydrate,dsstox_cid_928,acmc-1c7xm,dsstox_rid_75872,ni.so4.6h2o,dsstox_gsid_20928 PubChem CID: 5284429 ChEBI: CHEBI:53437 IUPAC-Name: Nickel(2+); Sulfat; Hexahydrat SMILES: O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
Nickel(II)-sulfat-Hexahydrat, 99.97 % min. (Metallbasis), Thermo Scientific Chemicals
CAS: 10101-97-0 Summenformel: H12NiO10S Molekulargewicht (g/mol): 262.839 MDL-Nummer: MFCD00149813 InChI-Schlüssel: RRIWRJBSCGCBID-UHFFFAOYSA-L Synonym: nickel sulfate hexahydrate,nickel ii sulfate hexahydrate,nickel 2+ sulfate hexahydrate,nickel ii sulfate,nickelsulfatehexahydrate,dsstox_cid_928,acmc-1c7xm,dsstox_rid_75872,ni.so4.6h2o,dsstox_gsid_20928 PubChem CID: 5284429 ChEBI: CHEBI:53437 IUPAC-Name: Nickel(2+); Sulfat; Hexahydrat SMILES: O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
Nickel(II)-Sulfat-Hexahydrat, reinst, SLR, Fisher Chemical
CAS: 10101-97-0 Summenformel: H12NiO10S Molekulargewicht (g/mol): 262.839 MDL-Nummer: 149813 InChI-Schlüssel: RRIWRJBSCGCBID-UHFFFAOYSA-L Synonym: nickel sulfate hexahydrate,nickel ii sulfate hexahydrate,nickel 2+ sulfate hexahydrate,nickel ii sulfate,nickelsulfatehexahydrate,dsstox_cid_928,acmc-1c7xm,dsstox_rid_75872,ni.so4.6h2o,dsstox_gsid_20928 PubChem CID: 5284429 ChEBI: CHEBI:53437 IUPAC-Name: Nickel(2+); Sulfat; Hexahydrat SMILES: O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
Nickel(II)-sulfat-Hexahydrat, Reagenzgütegrad, Honeywell™
CAS: 10101-97-0 Summenformel: H12NiO10S Molekulargewicht (g/mol): 262.839 MDL-Nummer: MFCD00149813 InChI-Schlüssel: RRIWRJBSCGCBID-UHFFFAOYSA-L Synonym: Nickelsulfat-Hexahydrat,Nickel-II-sulfat-Hexahydrat,Nickel2+-Sulfat-Hexahydrat,Nickel-II-sulfat,nickelsulfatehexahydrate,dsstox_cid_928,acmc-1c7xm,dsstox_rid_75872,ni.so4.6h2o,dsstox_gsid_20928 PubChem CID: 5284429 ChEBI: CHEBI:53437 IUPAC-Name: Nickel(2+); Sulfat; Hexahydrat SMILES: O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
Nickelsulfat, 6 -Hydrat, kristallin, BAKER ANALYZED™ A.C.S. Reagenz, J.T.Baker™
CAS: 10101-97-0 Summenformel: H12NiO10S Molekulargewicht (g/mol): 262.839 InChI-Schlüssel: RRIWRJBSCGCBID-UHFFFAOYSA-L Synonym: Nickelsulfat-Hexahydrat,Nickel-II-sulfat-Hexahydrat,Nickel2+-Sulfat-Hexahydrat,Nickel-II-sulfat,nickelsulfatehexahydrate,dsstox_cid_928,acmc-1c7xm,dsstox_rid_75872,ni.so4.6h2o,dsstox_gsid_20928 PubChem CID: 5284429 ChEBI: CHEBI:53437 IUPAC-Name: Nickel(2+); Sulfat; Hexahydrat SMILES: O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
Nickel(II)-sulfat-Hexahydrat, ≥99.99% Spurenmetallbasis
CAS: 10101-97-0 Summenformel: H12NiO10S Molekulargewicht (g/mol): 262.839 MDL-Nummer: MFCD00149813 InChI-Schlüssel: RRIWRJBSCGCBID-UHFFFAOYSA-L Synonym: Nickelsulfat-Hexahydrat,Nickel-II-sulfat-Hexahydrat,Nickel2+-Sulfat-Hexahydrat,Nickel-II-sulfat,nickelsulfatehexahydrate,dsstox_cid_928,acmc-1c7xm,dsstox_rid_75872,ni.so4.6h2o,dsstox_gsid_20928 PubChem CID: 5284429 ChEBI: CHEBI:53437 IUPAC-Name: Nickel(2+); Sulfat; Hexahydrat SMILES: O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
Nickel(II)-sulfat-Hexahydrat, ACS-Reagenz, Honeywell Fluka™
CAS: 10101-97-0 Summenformel: H12NiO10S Molekulargewicht (g/mol): 262.839 MDL-Nummer: MFCD00149813 InChI-Schlüssel: RRIWRJBSCGCBID-UHFFFAOYSA-L Synonym: Nickelsulfat-Hexahydrat,Nickel-II-sulfat-Hexahydrat,Nickel2+-Sulfat-Hexahydrat,Nickel-II-sulfat,nickelsulfatehexahydrate,dsstox_cid_928,acmc-1c7xm,dsstox_rid_75872,ni.so4.6h2o,dsstox_gsid_20928 PubChem CID: 5284429 ChEBI: CHEBI:53437 IUPAC-Name: Nickel(2+); Sulfat; Hexahydrat SMILES: O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]
Nickel(II)-sulfat-Hexahydrat, Puriss, p.a., ACS-Reagenz, 99 -102.0 %, Honeywell Fluka™
CAS: 10101-97-0 Summenformel: H12NiO10S Molekulargewicht (g/mol): 262.839 MDL-Nummer: MFCD00149813 InChI-Schlüssel: RRIWRJBSCGCBID-UHFFFAOYSA-L Synonym: Nickelsulfat-Hexahydrat,Nickel-II-sulfat-Hexahydrat,Nickel2+-Sulfat-Hexahydrat,Nickel-II-sulfat,nickelsulfatehexahydrate,dsstox_cid_928,acmc-1c7xm,dsstox_rid_75872,ni.so4.6h2o,dsstox_gsid_20928 PubChem CID: 5284429 ChEBI: CHEBI:53437 IUPAC-Name: Nickel(2+); Sulfat; Hexahydrat SMILES: O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Ni+2]