missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF711 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25166-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CDK4 is a member of the Ser/Thr protein kinase family. This protein is highly similar to the gene products of S. cerevisiae cdc28 and S. pombe cdc2. It is a catalytic subunit of the protein kinase complex that is important for cell cycle G1 phase progression. The activity of this kinase is restricted to the G1-S phase, which is controlled by the regulatory subunits D-type cyclins and CDK inhibitor p16(INK4a). This kinase was shown to be responsible for the phosphorylation of retinoblastoma gene product (Rb). Mutations in this gene as well as in its related proteins including D-type cyclins, p16(INK4a) and Rb were all found to be associated with tumorigenesis of a variety of cancers. Multiple polyadenylation sites of the gene have been reported.Spezifikation
ZNF711 | |
Polyclonal | |
Unconjugated | |
ZNF711 | |
CMPX1, Zfp711, Zinc finger protein 6, zinc finger protein 711, ZNF4, ZNF5, ZNF6, ZNF711 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
245595, 7552 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A2ANX9, Q9Y462 | |
Zfp711, ZNF711 | |
ZNF711 Fusion Protein Ag18309 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |