missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water, Sterile, Carl Roth™
Click to view available options
Menge:
1 L
Beschreibung
- Empirical formula H2O
- Molar mass (M) 18,02 g/mol
- Density (D) 1,0
- Boiling point (bp) 100 °C
- Melting point (mp)0 °C
- CAS-Nr. [7732-18-5]
- EG-Nr. 231-791-2
- Pyrogenity (endotoxins): <0.25 E.U./ml
- Meets in all aspects the demands of Ph.Eur. for Aqua purificata (purified water)
Spezifikation
Spezifikation
Menge | 1 L |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'