Learn More
KARTELL Volumetric Flask
Volumetric Flask, made of transparent fluorinated resin, has high chemical and temperature resistance.
Marke: KARTELL 0169900
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Volumetric flask made of transparent fluorinated resin with high chemical and temperature resistance (-250°C to +270°C).
- Supplied with standard DIN stoppers in PFA.
- Individually calibrated ringmark according to DIN EN ISO 1042, class “A”, Standards.
- Laser engraved lot number.
Spezifikation
500 mL | |
Nicht autoklavierbar | |
No | |
Unsteril | |
Unbehandelt |
PFA | |
Reusable | |
Enghals | |
Flach |