Learn More
Cole-Parmer Thermoplastic Elastomer (TPE) Tubing
Alle ansehen Cole-Parmer Produkte
Click to view available options
Innendurchmesser (angloamerikanisch):
1/16 in.
1/2 in.
1/32 in.
1/4 in.
1/8 in.
1 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/8 in.
Außendurchmesser (angloamerikanisch):
1/2 in.
1/4 in.
1/8 in.
1 1/4 in.
1 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
7/16 in.
7/32 in.
7/8 in.
9/16 in.
Packungsgröße:
15 Meter
Beschreibung
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. E in., cellent for use in peristaltic pump applications. Great alternative to silicone tubing. Does not contain PVC, DEHP, phthalates, or other plasticizers. Ultra-low e in., tractables and leachables. Low gas and o in., ygen permeability. Low protein binding and nonpyrogenic material. Thermally weldable. USP Class VI, USP 661, and RoHS compliant and BPOG tested
Spezifikation
Spezifikation
| Zertifizierung/Konformität | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Innendurchmesser (angloamerikanisch) | 3/4 in. |
| Material | TPE |
| Außendurchmesser (angloamerikanisch) | 1 in. |
| Gesamtlänge (angloamerikanisch) | 50 ft. |
| Gesamtlänge (metrisch) | 50 ft. |
| Sterilisationsmethode | 135°C |
| Farbe | Clear |