Learn More
TGF-beta Antibody (TGFB/510), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP3-08739PE
Artikelnummer. 18608797
  • 736.00 € / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
TGF-beta Monoclonal antibody specifically detects TGF-beta in Human, Mouse, Rat, Bovine, Canine, Hamster, Monkey samples. It is validated for ELISA, Functional Assay
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
TGF-beta
Monoclonal
PE
CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Full length recombinant human TGF-beta protein
0.1 mL
Cytokine Research, Extracellular Matrix
7040
Store at 4°C in the dark.
IgG1 κ
ELISA, Functional Assay
TGFB/510
PBS
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse, Rat, Bovine, Canine