Learn More
Tuttnauer Temperature Hold Cycle
Special program with programmable holding temperature at the end of the cycle. This will prevent that media will cool down completely. Must specify the cycle parameters, the holding temperature after the end of sterilization and the length of time to hold the temperature.
Marke:  Tuttnauer ADD222-0460
Artikelnummer. 17829538
  • 1680.00 € / 1 Stück
Verfügbar ab: 21-05-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
  • Special program with programmable holding temperature at the end of the cycle.
  • This will prevent that media will cool down completely.
  • Must specify the cycle parameters, the holding temperature after the end of sterilization and the length of time to hold the temperature.
Spezifikation
Spezifikation
Software
Dokumentation
One moment while we fetch your results.
Name des Produkts
Tuttnauer Temperature Hold Cycle > Temperature Hold Cycle