missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptococcus pneumoniae Monoclonal Antibody (F76G)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA518264
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody is reactive with the S. pneumoniae common C-polysaccharide (backbone epitope). Purity is >90%. This antibody is recommended as a detection antibody in ELISA applications (conjugated).
There are more than 85 antigenic types of S. pneumoniae, which are determined by capsule antigens. Altough S. pneumoniae is part of the normal respiratory flora, invasion commonly results in pneumonia as well as meningitis. S. pneumoniae is protected from phagocytosis by its polysaccharide capsule which is also a prominent virulence factor. Infections can usually be treated with antibiotics, although resistant strains are emerging.
Spezifikation
| Streptococcus pneumoniae | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG3 |
| ELISA, Immunocytochemistry | |
| F76G | |
| Unconjugated | |
| Pneumococcus; s pneumoniae; S. pneumoniae | |
| Strep pneumoniae native protein. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur