missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPPL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543867
This item is not returnable.
View return policy
Description
Peptide sequence: VMVKVATQPA DNPLDVLSRK LHLGPNVGRD VPRLSLPGKL VFPSSTGSHF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
SPPL3 Polyclonal antibody specifically detects SPPL3 in Human samples. It is validated for Western Blot.Specifications
SPPL3 | |
Polyclonal | |
Unconjugated | |
SPPL3 | |
4833416I09Rik; fb94d08; Imp2; IMP-2; Intramembrane protease 2; MDHV1887; Presenilin homologous protein 1; presenilin-like protein 4; PRO4332; PSH1; PSL4; putative intramembrane aspartyl protease; RGD1562826; signal peptide peptidase 3; signal peptide peptidase like 3; signal peptide peptidase-like 3; signal peptide peptidase-like 3, pseudogene 1; Signal peptide peptidase-like 3-like protein; signal peptide peptidase-like protein 3; Sppl3; sppl3 protein; Sppl3-ps1; SPP-like 3; SPP-like 3 protein; upregulated during skeletal muscle growth 3; Usmg3; wu:fb94d08; zgc:114093 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
121665 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TCT6 | |
SPPL3 | |
Synthetic peptide directed towards the middle region of human UNQ1887 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |