Learn More
SP Bel-Art™ SP Bel-Art Proculture Cryotube Floating Bubble Rack
Fits most cryotubes and holds them tightly in a circular pattern formed by 12 outer and 8 inner openings
Marke: SP Bel-Art™ F18876-0020
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Excellent for holding cryotubes during work or incubation
- Floats in water; can also be used for quickly thawing frozen cells in a 37°C water bath
- Easily remove tubes by holding the center handle and pushing the rack down against the bench until the legs touch
- Dimensions: 6.4 mm (0.24 in.) thick; 95 mm (3.75 in) in diameter; 19 mm (0.75 in.) long legs
- Autoclavable
Spezifikation
Polypropylen | |
20 | |
Je 1 |