Learn More
SP Bel-Art™ SP Bel-Art Proculture Cryotube Floating Bubble Rack
Fits most cryotubes and holds them tightly in a circular pattern formed by 12 outer and 8 inner openings
Marke:  SP Bel-Art™ F18876-0020
Artikelnummer. 17499793
  • 36.15 € / 1 Stück
Verfügbar ab: 21-11-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
  • Excellent for holding cryotubes during work or incubation
  • Floats in water; can also be used for quickly thawing frozen cells in a 37°C water bath
  • Easily remove tubes by holding the center handle and pushing the rack down against the bench until the legs touch
  • Dimensions: 6.4 mm (0.24 in.) thick; 95 mm (3.75 in) in diameter; 19 mm (0.75 in.) long legs
  • Autoclavable
Spezifikation
Spezifikation
Polypropylen
20