missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ryanodine, Tocris Bioscience™
Click to view available options
Menge:
1 mg
Beschreibung
Potent inhibitor of Ca2+ release from sarcoplasmic reticulum (IC50 values are 10 and 2.8 nM in skeletal and cardiac muscle respectively).
Spezifikation
Spezifikation
Chemischer Name oder Material | Ryanodine |
CAS | 15662-33-6 |
Summenformel | C25H35NO9 |
Menge | 1 mg |
Synonym | ryanodine |
InChI-Schlüssel | JJSYXNQGLHBRRK-FXKAWTGQSA-N |
SMILES | CC1CCC2(C3(CC4(C5(C(C(C3(C5(C2(C1O)O4)O)O)OC(=O)C6=CC=CN6)(C(C)C)O)C)O)C)O |
Molekulargewicht (g/mol) | 493.553 |
PubChem CID | 122173044 |
Formelmasse | 493.55 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'