missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11005-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This is a rabbit polyclonal antibody raised against the C terminus of human RPL3.
Ribosomes, the complexes that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L3P family of ribosomal proteins and it is located in the cytoplasm. The protein can bind to the HIV-1 TAR mRNA, and it has been suggested that the protein contributes to tat-mediated transactivation. This gene is co-transcribed with several small nucleolar RNA genes, which are located in several of this gene′s introns. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.Spezifikation
RPL3 | |
Polyclonal | |
Unconjugated | |
RPL3 | |
60S ribosomal protein L3, ribosomal protein L3, RPL3, TARBP B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27367, 6122 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P27659, P39023 | |
Rpl3 | |
RPL3 Fusion Protein Ag1436 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |