Learn More
STEMCELL Technologies RosetteSep™ HLA T Cell Enrichment Cocktail
Artikelnummer. 17137671
Encompass
Click to view available options
Menge:
1000 ml Vollblut
250 ml Vollblut
Packungsgröße:
1 Set
Artikelnummer. 17137671
Marke: STEMCELL Technologies 15081HLA
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Immunodensity isolation of untouched T cells for HLA analysis
  • The RosetteSep™ HLA T Cell Enrichment Cocktail is designed to isolate T cells from whole blood by negative selection.
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes (TAC) recognizing non-T cells and red blood cells (RBCs).
  • When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (Catalog #15705) or Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs.
  • The purified T cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium.
  • The desired cells are immediately ready for serology or flow cytometry crossmatch assays, or other downstream applications.
Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled For Research Use Only. Please note there have been no changes to the form, function and quality assurances associated with these products.
Spezifikation
Zelltyp T Cells
Zur Verwendung mit (Anwendung) Cell Isolation; In Vitro Diagnostic
Menge 1000 ml Vollblut
Berichtigung von Produktinhalten
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.