Learn More
STEMCELL Technologies RoboSep™ Human FITC Positive Selection Kit II
Click to view available options
Menge:
1 x 109 Zellen
Packungsgröße:
1 Set
Beschreibung
- The EasySep™ Human FITC Positive Selection Kit II is designed to isolate cells that are labeled with FITC-conjugated antibodies
- Desired cells are targeted with antibody complexes recognizing FITC and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- This product replaces the EasySep™ Human FITC Positive Selection Kit (Catalog #18552)
Spezifikation
Spezifikation
| Zelltyp | B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells |
| Zur Verwendung mit (Anwendung) | Cell Isolation |
| Menge | 1 x 109 Zellen |