missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNA Polymerase II/POLR2A Antibody (POLR2A/9089R), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP3-24041PCP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
RNA Polymerase II/POLR2A Monoclonal antibody specifically detects RNA Polymerase II/POLR2A in Human, Yeast samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Spezifikation
RNA Polymerase II/POLR2A | |
Monoclonal | |
PerCP | |
DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2 | |
Ten repeats of synthetic peptide YSPTSPS using chemically synthesized phospho-Ser5 | |
0.1 mL | |
Chromatin Research, Epigenetics, Phospho Specific, Transcription Factors and Regulators | |
5430 | |
Store at 4°C in the dark. | |
IgG κ |
Western Blot, Immunohistochemistry (Paraffin) | |
POLR2A/9089R | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Yeast | |
Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
RNA Polymerase II/POLR2A Antibody (POLR2A/9089R), PerCP, Novus Biologicals™ > 0.1 mL, PerCP
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur