Learn More
Invitrogen™ RING1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA527476
Beschreibung
Recommended positive controls: 293T. Predicted reactivity: Mouse (98%), Rat (99%), Xenopus laevis (91%), Dog (99%), Pig (99%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene belongs to the RING finger family, members of which encode proteins characterized by a RING domain, a zinc-binding motif related to the zinc finger domain. The gene product can bind DNA and can act as a transcriptional repressor. It is associated with the multimeric polycomb group protein complex. The gene product interacts with the polycomb group proteins BMI1, EDR1, and CBX4, and colocalizes with these proteins in large nuclear domains. It interacts with the CBX4 protein via its glycine-rich C-terminal domain. The gene maps to the HLA class II region, where it is contiguous with the RING finger genes FABGL and HKE4.
Spezifikation
| RING1 | |
| Polyclonal | |
| Unconjugated | |
| RING1 | |
| E3 ubiquitin-protein ligase RING1; Polycomb complex protein RING1; really interesting new gene 1 protein; RING finger protein 1; RING1; Ring1A; RING-type E3 ubiquitin transferase RING1; Rnf1; Transcription repressor Ring1A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6015 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q06587 | |
| RING1 | |
| Recombinant protein encompassing a sequence within the center region of human RING1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.