missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig CALR / Calreticulin Protein
A DNA sequence encoding the Sus scrofa (Pig) CALR / Calreticulin, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP9027-ye-10ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
NP_001167604.1 | |
CALR / Calreticulin Protein | |
Research Use Only | |
Calr | |
Recombinant Protein | |
Yeast | |
EPTIYFKEQF LDGDGWTDRW IESKHKPDFG RFVLSSGKFY GDQEKDKGLQ TSQDARFYAL SARFEPFSNK GQTLVVQFTV KHEQNIDCGG GYVKLFPDGL DQTDMHGDSE YNIMFGPDIC GPGTKKVHVI FNYKGKNVLI NKDIRCKDDE FTHLYTLIVR PDNTYEVKID NSQVESGSLE DDWDFLPPKK IKDPDAVKPE DWDERAKIDD PTDSKPEDWD KPEHIPDPDA KKPEDWDEEM DGEWEPPVIQ NPEYKGEWKP RQIDNPDYKG TWIHPEIDNP EYSPDSNIYA YENFAVLGLD LWQVKSGTIF DNFLITNDEA YAEEFGNETW GVTKAAEKQM KDKQDEEQRL KEEEEEKKRK EEEEVDKEDE EDKDEDEEEE DEKEEEEEED AAAGQAKDEL | |
Greater than 90% as determined by SDS-PAGE. |
100381266 | |
10 μg | |
Not determined. | |
Full Length | |
Pig | |
His | |
Tris-based buffer, 50% glycerol |