missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TNF-alpha (aa 80-235) Protein
Click to view available options
Menge:
10 μg
Packungsgröße:
10 Mikrogramm
Beschreibung
The Recombinant Mouse TNF-alpha (aa 80-235) Protein is derived from E. coli. The Recombinant Mouse TNF-alpha (aa 80-235) Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
| Zusammensetzung | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gen-ID (Entrez) | 21926 |
| Molekulargewicht | 17 kDa |
| Name | TNF-alpha aa 80-235 |
| Menge | 10 μg |
| Quelle | E. coli-derived mouse TNF-alpha protein Leu80-Leu235,with an N-terminal Met |
| Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Strukturform | A small amount of recombinant protein lacking the N-terminal methionine and four additional amino acid residues may be also present. |
| Gen-Alias | APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha |
| Konjugat | Unconjugated |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur