missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TLR3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marke: R&D Systems™ 3005-TR-050
265.22 EUR gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Mouse TLR3 Protein is derived from NS0. The Recombinant Mouse TLR3 Protein has been validated for the following applications: Bioactivity.Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS. | |
77.7 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
142980 | |
TLR3 | |
Mouse myeloma cell line,NS0-derived mouse TLR3 protein Cys29-Leu705,with a C-terminal 6-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |