missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Cadherin-4/R-Cadherin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marke: R&D Systems™ 6677-CA-050
285.09 EUR gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Mouse Cadherin-4/R-Cadherin Protein is derived from NS0. The Recombinant Mouse Cadherin-4/R-Cadherin Protein has been validated for the following applications: Bioactivity.Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS. | |
78.5 & 62.5 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
12561 | |
Cadherin-4/R-Cadherin | |
Mouse myeloma cell line,NS0-derived mouse Cadherin-4/R-Cadherin protein Met1-Ala731 with a C-terminal 6-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |