missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Vascular endothelial growth factor D Protein
A DNA sequence encoding the Homo sapiens (Human) Vascular endothelial growth factor D, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8578-ec-200ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
NP_004460.1 | |
Vascular endothelial growth factor D Protein | |
Research Use Only | |
VEGFD | |
Recombinant Protein | |
E. coli | |
FAATFYDIET LKVIDEEWQR TQCSPRETCV EVASELGKST NTFFKPPCVN VFRCGGCCNE ESLICMNTST SYISKQLFEI SVPLTSVPEL VPVKVANHTG CKCLPTAPRH PYSIIRR | |
Greater than 90% as determined by SDS-PAGE. |
2277 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |