missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human R-Spondin 4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke: R&D Systems™ 4575-RS-025/CF
311.01 EUR gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Human R-Spondin 4 Protein is derived from CHO. The Recombinant Human R-Spondin 4 Protein has been validated for the following applications: Bioactivity.Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS. | |
24.1 kDa (monomer) | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
343637 | |
R-Spondin 4 | |
Chinese Hamster Ovary cell line,CHO-derived human R-Spondin 4 protein Met1-Pro234 | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |