missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-10 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke: R&D Systems™ 217-IL-025
964.12 EUR gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Measured in a cell proliferation assay using MC/9-2 mouse mast cells. Thompson-Snipes, L. et al. (1991) J. Exp. Med. 173:507. The ED50 for this effect is 0.075-0.750 ng/mL.Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
18.6 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
Recombinant |
3586 | |
IL-10 | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-10 protein Ser19-Asn178 | |
Noncovalently-linked homodimer | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |