missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Homeobox protein TGIF2LX Protein
A DNA sequence encoding the Homo sapiens (Human) Homeobox protein TGIF2LX, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP7806-ec-50ug
Weitere Details : Netto-Gewicht : 0.01000kg
Spezifikation
NP_620410.3 | |
Homeobox protein TGIF2LX Protein | |
Research Use Only | |
TGIF2LX | |
Recombinant Protein | |
E. coli | |
MEAAADGPAE TQSPVEKDSP AKTQSPAQDT SIMSRNNADT GRVLALPEHK KKRKGNLPAE SVKILRDWMY KHRFKAYPSE EEKQMLSEKT NLSLLQISNW FINARRRILP DMLQQRRNDP IIGHKTGKDA HATHLQSTEA SVPAKSGPSG PDNVQSLPLW PLPKGQMSRE KQPDPESAPS QKLTGIAQPK KKVKVSVTSP SSPELVSPEE HADFSSFLLL VDAAVQRAAE LELEKKQEPN P | |
Greater than 90% as determined by SDS-PAGE. |
90316 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |