Learn More
enQuireBio™ Recombinant Human Golgi SNAP receptor complex member 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Golgi SNAP receptor complex member 2, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6107-ec-200ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
NP_001012529.1 | |
Golgi SNAP receptor complex member 2 Protein | |
Research Use Only | |
GOSR2 | |
Recombinant Protein | |
E. coli | |
MDPLFQQTHK QVHEIQSCMG RLETADKQSV HIVENEIQAS IDQIFSRLER LEILSSKEPP NKRQNARLRV DQLKYDVQHL QTALRNFQHR RHAREQQERQ REELLSRTFT TNDSDTTIPM DESLQFNSSL QKVHNGMDDL ILDGHNILDG LRTQRLTLKG TQKKILDIAN MLGLSNTVMR LIEKRAFQDK | |
Greater than 90% as determined by SDS-PAGE. |
9570 | |
200 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |