missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human EpCAM/TROP1 Fc Alexa Fluor™ 647 Protein
Marke: R&D Systems™ AFR960-020
431.95 EUR gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Measured by flow cytometry for its ability to bind Human EpCAM/TROP-1 Antibody conjugated fluorescent beads at 0.500-2.00 μg/mL (100 μL/well). Please Note: Optimal dilutions should be determined by each laboratory for each application.Spezifikation
CAA32870 | |
Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. | |
M.W. Observed: 60-70 kDa, under reducing conditions., M.W. Theoretical: 54 kDa | |
Mouse myeloma cell line, NS0-derived human EpCAM/TROP1 protein Human EpCAM (Gln24-Lys265) (N-terminus) Accession # CAA32870.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) | |
<1.0 EU per 1 μg of the protein by the LAL method. | |
Alexa Fluor 647 | |
EpCAM/TROP1 |
Bioactivity | |
4072 | |
20 μg | |
Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 3 months, -20 to -70 °C under sterile conditions after opening. | |
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2, GA733-2EGP34, gp40, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining. |