Learn More
R&D Systems™ Recombinant Human CCL4L1/MIP-1 beta Isoform LAG-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke: R&D Systems™ 3046-MB-025/CF
168.46 EUR gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Human CCL4L1/MIP-1 beta Isoform LAG-1 is derived from E. coli. The Recombinant Human CCL4L1/MIP-1 beta Isoform LAG-1 has been validated for the following applications: Bioactivity.Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS. | |
7.8 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
9560 | |
CCL4L1/LAG-1 | |
E. coli-derived human CCL4L1/LAG-1 protein Ala24-Asn92 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |