Learn More
R&D Systems™ Recombinant Human Angiogenin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Marke: R&D Systems™ 265-AN-250/CF
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Human Angiogenin Protein is derived from E. coli. The Recombinant Human Angiogenin Protein has been validated for the following applications: Enzyme Activity.Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS. | |
14 kDa | |
250μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
283 | |
Angiogenin | |
E. coli-derived human Angiogenin protein Gln25-Pro147 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |