Learn More
R&D Systems™ Recombinant Human Angiogenin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Marke:  R&D Systems™ 265-AN-250/CF
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 18188360
  • 1364.98 € / 250 Mikrogramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Entdecken Sie weitere Sonderangebote
Beschreibung
Beschreibung
The Recombinant Human Angiogenin Protein is derived from E. coli. The Recombinant Human Angiogenin Protein has been validated for the following applications: Enzyme Activity.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS.
14 kDa
250μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
283
Angiogenin
E. coli-derived human Angiogenin protein Gln25-Pro147
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.