Learn More
R&D Systems™ Recombinant Human ADAM12 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Marke:  R&D Systems™ 4416-AD-020
Artikelnummer. 18137580
  • 485.00 € / 20 Mikrogramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Entdecken Sie weitere Sonderangebote
Beschreibung
Beschreibung
The Recombinant Human ADAM12 Protein is derived from CHO. The Recombinant Human ADAM12 Protein has been validated for the following applications: Enzyme Activity.
Spezifikation
Spezifikation
Supplied as a 0.2 μm filtered solution in Sodium Acetate, CaCl 2 and NaCl.
55 kDa (Pro) & 34 kDa (Mature)
20 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
8038
ADAM12
Chinese Hamster Ovary cell line,CHO-derived human ADAM12 protein Arg29-Ser513,with an N-terminal signal peptide and a C-terminal 6-His tag
Pro & Mature. Recombinant Human ADAM12 is prone to proteolytic clevage at C-terminus. The predominant form of the purified protein lacks the His tag.
Recombinant
Produktvorschläge