Learn More
R&D Systems™ Recombinant Human ADAM12 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Marke: R&D Systems™ 4416-AD-020
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Human ADAM12 Protein is derived from CHO. The Recombinant Human ADAM12 Protein has been validated for the following applications: Enzyme Activity.Spezifikation
Supplied as a 0.2 μm filtered solution in Sodium Acetate, CaCl 2 and NaCl. | |
55 kDa (Pro) & 34 kDa (Mature) | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
8038 | |
ADAM12 | |
Chinese Hamster Ovary cell line,CHO-derived human ADAM12 protein Arg29-Ser513,with an N-terminal signal peptide and a C-terminal 6-His tag | |
Pro & Mature. Recombinant Human ADAM12 is prone to proteolytic clevage at C-terminus. The predominant form of the purified protein lacks the His tag. | |
Recombinant |
Berichtigung von Produktinhalten