missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Rat CXCL1 (GRO alpha) Recombinant Protein, PeproTech®

Artikelnummer. 17842933
Click to view available options
Menge:
100 μg
2 x 500 μg
25 μg
250 μg
5 μg
500 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
2 Stück
25 Mikrogramm
250 Mikrogramm
5 Mikrogramm
500 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 17842933

Marke: Gibco™ 40010500UG

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

400-10-1MG will be provided as 2 x 500 μg (400-10-500UG). Recombinant Rat GRO/KC is a 7.8 kDa protein consisting of 72 amino acids, including the 'ELR' motif common to the CXC chemokine family that binds to CXCR1 or CXCR2. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CXCL1, CXCL2, and CXCL3, also known respectively as GRO alpha, GRO beta (MIP-2 alpha) and GRO gamma (MIP-2 beta), are members of the CXC subfamily of chemokines. Mature CXCL1/2/3 proteins bind with high affinity to the IL-8 receptor type B and are potent neutrophil attractants and activators. In vitro, the processed forms GRO-alpha(4-73), GRO-alpha(5-73) and GRO-alpha(6-73) show a 30-fold higher chemotactic activity.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P14095
Zur Verwendung mit (Anwendung) Control, Western Blot Control, ELISA Standard, Bioactivity
Zusammensetzung Protein with no preservative
Gen-ID (Entrez) 81503
Molekulargewicht 7.8 kDa
Name Rat CXCL1 (GRO alpha)
Reinigungsverfahren Purified
Menge 500 μg
Quelle E. coli
Lagerungsbedingungen -20°C
Kennzeichnung RUO
Endotoxin-Konzentration <1 EU/μg
Gen-Alias alpha-chemokine; C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 1; chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha); Cinc1; CINC-1; CXC; CXC motif chemokine; C-X-C motif chemokine 1; C-X-C motif chemokine ligand 1; CXCL; CXCL1; CXCL-1; Cytokine-induced neutrophil chemoattractant 1; fibroblast secretory protein; Fsp; Gro; GRO a; GRO α; Gro1; GRO1 oncogene; GRO1 oncogene (melanoma growth stimulating activity, alpha); GRO1 oncogene (melanoma growth-stimulating activity); GROa; GROalpha; GRO-alpha(1-73); GRO-alpha(4-73); GRO-alpha(5-73); GRO-alpha(6-73); Growth-regulated alpha protein; GROα; Hematopoietic synergistic factor; HSF; KC; KC(5-72); KC/GR)-alpha; KC/GRO-alpha; KC-T; melanoma growth stimulating activity, alpha; Melanoma growth stimulatory activity; melanoma growth stimulatory activity alpha; MGSA; MGSA alpha; MGSA-a; N51; NAP3; NAP-3; Neutrophil-activating protein 3; platelet-derived growth factor-inducible protein KC; SCYB1; secretory protein N51
Gebräuchliche Bezeichnung CXCL1 (GRO alpha)
Gensymbol Cxcl1
Biologische Aktivität Determined by its ability to chemoattract rat neutrophils using a concentration range of 10.0-100.0 ng/ml
Produkttyp Protein
Konjugat Unconjugated
Rekombinant Recombinant
Sequenz APVANELRCQ CLQTVAGIHF KNIQSLKVMP PGPHCTQTEV IATLKNGREA CLDPEAPMVQ KIVQKMLKGV PK
Inhalt und Lagerung -20°C
Aktivität Determined by its ability to chemoattract rat neutrophils using a concentration range of 10.0-100.0 ng/ml.
Expressionssystem E. coli
Form Lyophilized
Reinheits- oder Qualitätsgrad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt