missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAP1A/RAP1B Recombinant Rabbit Monoclonal Antibody (JE35-61), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen MA550823
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Positive control: HeLa cell lysate, Jurkat cell lysate, NIH/3T3 cell lysate, HEK-293 cell lysate, HEK-293. Predicted band size: 20 kDa Subcellular Location: Cell membrane, Cytoplasm, Cytoplasm, perinuclear region, Cell junction, Early endosome; Cell membrane, Cytoplasm, cytosol, Cell junction.
RAP1A/RAP1B Recombinant Monoclonal antibody specifically detects RAP1A/RAP1B in Human, Mouse samples. It is validated for Flow Cytometry, Western BlotSpezifikation
RAP1A/RAP1B | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P61224, P62834, P62835, Q99JI6 | |
RAP1A, RAP1B | |
Synthetic peptide within Human RAP1A aa 93-142/184. | |
100 μL | |
Primary | |
Human, Mouse | |
Liquid |
Flow Cytometry, Western Blot | |
JE35-61 | |
Unconjugated | |
RAP1A | |
2810443E11Rik, AI848598, C21KG, G-22K, GTP-binding protein smg p21A, GTP-binding protein smg p21B, GTP-binding protein SMG-P21A, K-REV, KREV1, Krev-1, OK/SW-cl.11, RAL1B, rap 1A, RAP1, RAP1A, RAP-1A, RAP1A, member of RAS oncogene family, Rap1b, RAP1B, member of RAS oncogene family, Ras family small GTP binding protein RAP1B, RAS related protein 1b, RAS-related protein 1a, Ras-related protein Krev-1, Ras-related protein Rap-1A, RAS-related protein RAP1B, ras-related protein Rap-1b, RAS-related protein-1a, small GTP binding protein, SMGP21 | |
Rabbit | |
Protein A | |
RUO | |
109905, 215449, 5906, 5908 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |