missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit IgG control Rabbit, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech CL594-30000-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Rabbit IgG control Polyclonal antibody apecifically detetects Rabbit IgG control antigen in Samples. It is validated for Flow Cytometry.Spezifikation
Rabbit IgG control | |
Polyclonal | |
CoraLite 594 | |
Rabbit | |
Primary | |
Liquid |
Flow Cytometry | |
1000 ug/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
20 μL | |
Store at -20°C. Avoid exposure to light. | |
IgG |