Learn More
NPFFR2 Rabbit anti-Horse, Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NPFFR2.
Marke: Abnova PAB26577.50ug
Beschreibung
This gene encodes a member of a subfamily of G-protein-coupled neuropeptide receptors. This protein is activated by the neuropeptides A-18-amide (NPAF) and F-8-amide (NPFF) and may function in pain modulation and regulation of the opioid system. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Spezifikation
| NPFFR2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NPFFR2. | |
| In PBS (0.09% sodium azide) | |
| GPR74/NPFF2/NPGPR | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 10886 | |
| Equine, Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (16 ug/mL) The optimal working dilution should be determined by the end user. | |
| NPFFR2 | |
| NPFFR2 | |
| A synthetic peptide corresponding to 18 amino acids from 3rd extracellular domain of human NPFFR2. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only