missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quercetin, Tocris Bioscience™
Click to view available options
Menge:
100 mg
Beschreibung
Antitumor agent; induces apoptosis and inhibits synthesis of heat shock proteins. Inhibits many enzyme systems including tyrosine protein kinase, phospholipase A2, phosphodiesterases, mitochondrial ATPase, PI 3-kinase and protein kinase C. Can also activate Ca2+ and K+ channels and behaves as an agonist at estrogen (GPER) receptors. Blocks cellular entry by influenza A virus via an interaction with hemaglutinin. Inhibits SARS-CoV 3CLpro (IC50 = 73 μM).
Spezifikation
Spezifikation
Chemischer Name oder Material | Quercetin |
CAS | 117-39-5 |
Summenformel | C15H10O7 |
Menge | 100 mg |
Synonym | quercetin, sophoretin, meletin, xanthaurine, quercetine, quercetol, quercitin, quertine, 3,3',4',5,7-pentahydroxyflavone, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one |
InChI-Schlüssel | REFJWTPEDVJJIY-UHFFFAOYSA-N |
SMILES | C1=CC(=C(C=C1C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O)O)O |
IUPAC-Name | 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one |
Molekulargewicht (g/mol) | 302.238 |
PubChem CID | 5280343 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'