missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Propidium Iodide

Artikelnummer. 16412326
Encompass_Preferred
Click to view available options
Menge:
10 mg
50 mg
100 mg
500 mg
Packungsgröße:
10 Milligramm
50 Milligramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 16412326

Marke: MedChemExpress HYD0815

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

MedChemExpress Propidium Iodide is a red-fluorescent dye that can be used to stain cells.

Specifications

Inhalt und Lagerung 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Produkttyp Dye Reagents
Menge 500 mg
Versandbedingung Room temperature
Löslichkeit DMSO : 100 mg/mL (149.61 mM; Need ultrasonic) ∣ H2O : 3.57 mg/mL (5.34 mM; ultrasonic and warming and heat to 60°C)
Ziel HBV
Molekulargewicht 668.39 g/mol
CAS 25535-16-4
Synonym PI
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.