Learn More
PREB Mouse anti-Human, Unconjugated, Clone: 2F6A1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66981-1-IG-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The PREB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PREB. This antibody recognizes human antigen. The PREB antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spezifikation
PREB | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
2F6A1 | |
Unconjugated | |
20 μL | |
10113 | |
IgG1 |