Learn More
PP2A alpha/beta Recombinant Rabbit Monoclonal Antibody (SN70-08), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen MA532304
343.70 EUR gültig bis 2025-03-16
Benutzen Sie den Promotions-Code "24070" um Ihren Promotions-Preis zu erhalten.
Beschreibung
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes an alpha isoform of the catalytic subunit.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spezifikation
PP2A alpha/beta | |
Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide, pH 7.4 | |
P62714, P62715, P62716, P63330, P63331, P67775 | |
PPP2CA, PPP2CB | |
Recombinant protein within Human PP2A alpha + beta aa 260-309 | |
100 μL | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry, Western Blot, Western Blot | |
SN70-08 | |
Unconjugated | |
PPP2CB | |
AI115466, D8Ertd766e, fb17h07, fc05c02, fk24a01, hypothetical protein LOC550585, PP2A, Pp2a1, Pp2a2, PP2A-alpha, PP2Abeta, PP2A-beta, PP2Ac, PP2CA, PP2Calpha, PP2CB, ppp2ca, ppp2caa, PPP2CB, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform, protein phosphatase 2 catalytic subunit alpha, protein phosphatase 2 catalytic subunit alpha a, protein phosphatase 2 catalytic subunit beta, protein phosphatase 2, catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isozyme, protein phosphatase 2, catalytic subunit, beta isoform, protein phosphatase 2, catalytic subunit, beta isozyme, protein phosphatase 2,catalytic subunit alpha isozyme, protein phosphatase 2A alpha subunit, protein phosphatase 2A beta subunit, protein phosphatase 2A catalytic alpha subunit, protein phosphatase 2A catalytic subunit alpha isoform, protein phosphatase 2A catalytic subunit beta isoform, protein phosphatase 2A catalytic subunit, beta isoform, protein phosphatase 2a, catalytic subunit, alpha isoform, protein phosphatase 2A, catalytic subunit, beta isoform, protein phosphatase type 2A catalytic subunit, protein phosphatase-2A-alpha, protein phosphatase-2A-beta, R75353, replication protein C, RP C, RP-C, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform, serine/threonine protein phosphatase 2A, catalytic subunit, beta isoform, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform, Serine/threonine-protein phosphatase 2A catalytic subunit beta isoform, testicular tissue protein Li 146, wu:fb17h07, wu:fc05c02, wu:fk24a01, zgc:110641, zgc:55808, zgc:77172 | |
Rabbit | |
Protein A | |
RUO | |
19052, 19053, 24672, 24673, 406582, 550585, 5515, 5516 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.