Learn More
PIGO Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531201
338.68 EUR gültig bis 2025-03-29
Benutzen Sie den Promotions-Code "24111" um Ihren Promotions-Preis zu erhalten.
Benachrichtigungen:
Um den Rabatt von 33.33 % zu erhalten, müssen Kunden drei Einheiten desselben Produkts zum Listenpreis in einer einzigen Bestellung kaufen. Es gibt keine Begrenzung für die Anzahl von 3er Sets, die Kunden kaufen können. PCODE: 24111
Beschreibung
Recommended positive controls: A549, HeLa, HCT116. Predicted reactivity: Mouse (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid which contains three mannose molecules in its core backbone. The GPI-anchor is found on many blood cells and serves to anchor proteins to the cell surface. This protein is involved in the transfer of ethanolaminephosphate (EtNP) to the third mannose in GPI. At least two alternatively spliced transcripts encoding distinct isoforms have been found for this gene.
Spezifikation
PIGO | |
Polyclonal | |
Unconjugated | |
PIGO | |
GPI ethanolamine phosphate transferase 3; HPMRS2; phosphatidylinositol glycan anchor biosynthesis class O; phosphatidylinositol glycan anchor biosynthesis, class O; phosphatidylinositol glycan, class O; phosphatidylinositol-glycan biosynthesis class O protein; Pigo; PIG-O; RP11-182N22.4; UNQ632/PRO1249 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84720 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.97 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8TEQ8 | |
PIGO | |
Recombinant fragment corresponding to a region within amino acids 201 and 421 of Human PIGO | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.