Learn More
Gilac™ Picking Bin
Click to view available options
Farbe:
Blau
Gelb
Grau
Grün
Rot
Weiß
Packungsgröße:
1 Stück
Description
- Open fronted picking bins.
- Contents easily accessible.
- Stackable up to 2 metres.
- Stability reinforced through bin feet interlocking.
- Available in 6 colors, making it easier to differentiate the contents.
Specifications
Specifications
| Typ | Storage Bin |
| Material | HDPE |
| Farbe | Grau |
| Form | Rechteckig |