Learn More
WTW™ PhotoLab™ Color and PhotoLab™ Spectral Software
WTW™ PhotoLab™ Color and Data Spectral Software for the 7000/6000 Series photometers consists of two individual modules offering powerful photometric color measurement and GLP-compliant data management.
Marke: WTW™ 902763
Weitere Details : Netto-Gewicht : 0.12500kg
Beschreibung
- PC-triggered color measurement and analysis
- 'photoLab™ color' offers color measurement instead of color perception
- 'photoLab™ Data spectral' provides data management and analysis for the photoLab™ 7000/6000 Series
- Intuitive user interfaces
- Easy data management and outputs
- GLP-compliant
Spezifikation
GLP compliant | |
Photometric color measurement and data management | |
WTW™ Photometers 7000/6000 Series | |
Windows 7, Windows 8, Windows 10 |