missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipase C beta 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 26551-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The protein encoded by this gene catalyzes the formation of inositol 1,4,5-trisphosphate and diacylglycerol from phosphatidylinositol 4,5-bisphosphate. This reaction uses calcium as a cofactor and plays an important role in the intracellular transduction of many extracellular signals. This gene is activated by two G-protein alpha subunits, alpha-q and alpha-11. Two transcript variants encoding different isoforms have been found for this gene.Spezifikation
Phospholipase C beta 1 | |
Polyclonal | |
Unconjugated | |
PLCB1 | |
KIAA0581, Phospholipase C beta 1, Phospholipase C I, PI PLC, PLC 154, PLC beta 1, PLC I, PLC154, PLCB1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18795, 23236, 24654 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P10687, Q9NQ66, Q9Z1B3 | |
Plcb1 | |
Phospholipase C beta 1 Fusion Protein Ag24750 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Phospholipase C beta 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur