missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-HEC1 (Ser55) Recombinant Rabbit Monoclonal Antibody (HL3190)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA556735
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Phospho-HEC1 (Ser55) Recombinant Monoclonal Antibody for Western Blot
Hec1 is one of several proteins involved in spindle checkpoint signaling. This surveillance mechanism assures correct segregation of chromosomes during cell division by detecting unaligned chromosomes and causing prometaphase arrest until the proper bipolar attachment of chromosomes is achieved.
Spezifikation
| Phospho-HEC1 (Ser55) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O14777 | |
| NDC80 | |
| Carrier-protein conjugated synthetic peptide surrounding phospho Ser55 of human Hec1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL3190 | |
| Unconjugated | |
| NDC80 | |
| 2610020P18Rik; AV139730; HEC; HEC1; HEC-1; HEC2; HEC4; Highly expressed in cancer protein; highly expressed in cancer, rich in leucine heptad repeats; HsHec1; hsNDC80; kinetochore associated 2; kinetochore protein Hec1; Kinetochore protein NDC80 homolog; kinetochore-associated protein 2; KNTC2; NDC80; NDC80 homolog, kinetochore complex component; NDC80 homolog, kinetochore complex component (S. cerevisiae); NDC80 kinetochore complex component; NDC80 kinetochore complex component homolog; NDC80, kinetochore complex component; putative kinetochore protein NDC80 like protein; retinoblastoma-associated protein HEC; TID3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10403 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur